Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168370.1 | NP_001161842.1 | 24 | 4892 | ||
NM_014780.4 | NP_055595.2 | 24 | 4885 | |||
XM_005249503.2 | XP_005249560.1 | 24 | 4748 | |||
XM_006715285.1 | XP_006715348.1 | 24 | 4986 | |||
XM_011515019.2 | XP_011513321.1 | 24 | 4843 | |||
XM_011515020.2 | XP_011513322.1 | 24 | 4746 | |||
XM_011515021.1 | XP_011513323.1 | 19 | 2927 | |||
XM_017011533.1 | XP_016867022.1 | 24 | 4871 | |||
XM_017011534.1 | XP_016867023.1 | 24 | 4870 | |||
XM_017011535.1 | XP_016867024.1 | 24 | 4774 | |||
XM_017011536.1 | XP_016867025.1 | 24 | 5013 | |||
XM_017011537.1 | XP_016867026.1 | 24 | 4986 | |||
XM_017011538.1 | XP_016867027.1 | 24 | 4917 | |||
XM_017011539.1 | XP_016867028.1 | 24 | 4890 | |||
GenBank mRNA | AK292806.1 | - | 24 | 4822 | ||
AK302668.1 | - | 24 | 4829 | |||
AK307936.1 | - | 14 | 2313 | |||
BC033647.1 | - | 24 | 4867 | 5580027 | ||
D38548.1 | - | 24 | 4639 |