Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009624.1 | NP_001009624.1 | 3 | 150 | ||
GenBank mRNA | BC088264.1 | - | 3 | 150 | 7377141 | |
FQ220730.1 | - | 3 | 149 | |||
FQ229832.1 | - | 3 | 149 |