Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aknad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044303.2 | NP_001037768.2 | 3 | 917 | ||
XM_008761422.1 | XP_008759644.1 | 3 | 2098 | |||
GenBank mRNA | BC079051.1 | - | 3 | 917 | 7115381 |