Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006443.2 | NP_006434.1 | 3 | 345 | ||
NM_199184.1 | NP_954653.1 | 3 | 345 | |||
GenBank mRNA | AF040105.1 | - | 3 | 345 | ||
BC011683.2 | - | 3 | 339 | 3633266 | ||
BM747628.1 | - | 3 | 353 | |||
EU585603.1 | - | 3 | 353 |