Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDB3 SIRNA Assays ›
Gene Symbol:
LDB3
Gene Name:
LIM domain binding 3
Gene Aliases:
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP
Chromosome Location:
Chr.10: 86664785 - 86736072 on Build GRCh38
UniGene ID:
Hs.657271
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s195088
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080114.1
NP_001073583.1
2, 3
264
NM_001080115.1
NP_001073584.1
2, 3
264
NM_001080116.1
NP_001073585.1
2, 3
369
NM_001171610.1
NP_001165081.1
3, 4
386
NM_001171611.1
NP_001165082.1
2, 3
264
NM_007078.2
NP_009009.1
2, 3
264
XM_005269464.4
XP_005269521.1
3, 4
433
XM_005269466.4
XP_005269523.1
3, 4
432
XM_005269468.4
XP_005269525.1
3, 4
429
XM_011539184.2
XP_011537486.1
3, 4
434
XM_011539185.2
XP_011537487.1
3, 4
2328
XM_011539186.2
XP_011537488.1
3, 4
434
XM_011539187.2
XP_011537489.1
3, 4
433
XM_011539188.2
XP_011537490.1
3, 4
434
XM_011539189.1
XP_011537491.1
3, 4
328
XM_011539190.1
XP_011537492.1
2, 3
264
XM_011539191.2
XP_011537493.1
3, 4
433
XM_011539193.1
XP_011537495.1
3, 4
328
XM_011539194.1
XP_011537496.1
3, 4
328
XM_011539195.2
XP_011537497.1
3, 4
430
XM_017015606.1
XP_016871095.1
3, 4
2328
XM_017015608.1
XP_016871097.1
3, 4
2328
XM_017015609.1
XP_016871098.1
3, 4
328
GenBank mRNA
AB014513.1
-
2, 3
262
AF276807.1
-
2, 3
241
AF276808.1
-
2, 3
241
AF276809.1
-
2, 3
241
AJ133766.1
-
2, 3
320
AJ133767.1
-
2, 3
241
AJ133768.1
-
2, 3
241
AK294696.1
-
3, 4
321
AK297871.1
-
3, 4
220
AK300692.1
-
3, 4
277
AK304760.1
-
3, 4
328
AL601484.1
-
2, 3
276
BC010929.2
-
2, 3
266
DA898241.1
-
2, 3
369
DC357269.1
-
3, 4
386
KF772970.1
-
2, 3
241
KU178543.1
-
2, 3
241
KU178544.1
-
2, 3
241
KU178545.1
-
2, 3
241
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
sarcomere organization
protein kinase C binding
protein binding
cytoskeletal protein binding
zinc ion binding
muscle alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-bqghl:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0