Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 2 | 216 | ||
NR_027409.1 | 11 | 1387 | ||||
XM_006720440.2 | XP_006720503.1 | 2 | 184 | |||
XM_017022018.1 | XP_016877507.1 | 9 | 1168 | |||
XM_017022019.1 | XP_016877508.1 | 8 | 864 | |||
GenBank mRNA | AK123221.1 | - | 7 | 732 | ||
BC032774.1 | - | 2 | 118 | |||
BC093673.1 | - | 2 | 162 | 7939518 | ||
BC104800.1 | - | 2 | 162 | 8143803 | ||
BX648160.1 | - | 10 | 1342 | |||
BX648575.1 | - | 10 | 1094 | |||
CR749361.1 | - | 3 | 278 |
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 2 | 216 | ||
NR_027410.1 | 11 | 1373 | ||||
XM_011521596.1 | XP_011519898.1 | 9 | 1061 | |||
XM_011521597.1 | XP_011519899.1 | 8 | 1012 | |||
XM_017022210.1 | XP_016877699.1 | 10 | 1365 | |||
XM_017022211.1 | XP_016877700.1 | 10 | 1134 | |||
XM_017022212.1 | XP_016877701.1 | 9 | 1688 | |||
XM_017022213.1 | XP_016877702.1 | 9 | 1090 | |||
XM_017022214.1 | XP_016877703.1 | 10 | 1365 | |||
XM_017022215.1 | XP_016877704.1 | 10 | 1365 | |||
XM_017022216.1 | XP_016877705.1 | 10 | 1394 | |||
XM_017022217.1 | XP_016877706.1 | 9 | 1316 | |||
XM_017022218.1 | XP_016877707.1 | 10 | 1992 | |||
GenBank mRNA | AK097551.1 | - | 2 | 205 | ||
BC143254.1 | - | 2 | 162 | 9051756 | ||
BX647673.1 | - | 10 | 1342 |