Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC146853.1 | - | 3, 4 | 271 | 9056781 |
Gene Symbol: RBMY1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144422.1 | - | 3, 4 | 271 | 9052943 |
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303410.1 | NP_001290339.1 | 3, 4 | 318 | ||
NM_152585.2 | NP_689798.1 | 3, 4 | 318 | |||
XM_017030033.1 | XP_016885522.1 | 2, 3 | 134 | |||
GenBank mRNA | BC030018.2 | - | 3, 4 | 318 | 5269012 | |
X76060.1 | - | 3, 4 | 300 |
Gene Symbol: RBMY1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006117.3 | NP_001006117.2 | 3, 4 | 383 | ||
XM_011531500.1 | XP_011529802.1 | 3, 4 | 254 | |||
XM_011531502.1 | XP_011529804.1 | 3, 4 | 254 | |||
XM_017030059.1 | XP_016885548.1 | 2, 3 | 134 | |||
GenBank mRNA | AK313597.1 | - | 3, 4 | 387 | ||
BC144577.1 | - | 3, 4 | 271 | 9053106 | ||
BC152392.1 | - | 2, 3 | 228 | 8860211 |