Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015022.1 | XP_016870511.1 | 6, 7 | 678 | ||
XM_017015024.1 | XP_016870513.1 | 6, 7 | 678 | |||
XM_017015025.1 | XP_016870514.1 | 6, 7 | 678 | |||
XM_017015033.1 | XP_016870522.1 | 2, 3 | 189 | |||
XM_017015036.1 | XP_016870525.1 | 7, 8 | 738 | |||
GenBank mRNA | AK293173.1 | - | 7, 8 | 673 | ||
BC069006.1 | - | 8, 9 | 783 |
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251514.1 | XP_005251571.1 | 7, 8 | 678 | ||
XM_005251515.1 | XP_005251572.1 | 7, 8 | 678 | |||
XM_011517964.2 | XP_011516266.1 | 7, 8 | 678 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263602.2 | XP_005263659.1 | 7, 8 | 750 | ||
XM_005263604.2 | XP_005263661.1 | 7, 8 | 750 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005277639.2 | XP_005277696.1 | 7, 8 | 738 | ||
XM_005277640.2 | XP_005277697.1 | 7, 8 | 738 | |||
XM_017014730.1 | XP_016870219.1 | 6, 7 | 678 | |||
XM_017014732.1 | XP_016870221.1 | 2, 3 | 189 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 7, 8 | 715 | ||
XM_005272751.1 | XP_005272808.1 | 7, 8 | 675 | |||
XM_017014455.1 | XP_016869944.1 | 7, 8 | 675 | |||
XM_017014457.1 | XP_016869946.1 | 8, 9 | 606 | |||
XM_017014460.1 | XP_016869949.1 | 7, 8 | 675 | |||
XM_017014461.1 | XP_016869950.1 | 9, 10 | 854 | |||
GenBank mRNA | BC082271.1 | - | 7, 8 | 611 |