Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005765.2 | NP_005756.2 | 2 | 187 | ||
GenBank mRNA | AF109363.1 | - | 2 | 148 | ||
AF248966.1 | - | 2 | 192 | |||
AF291814.1 | - | 2 | 174 | |||
AK075382.1 | - | 2 | 245 | |||
AK293510.1 | - | 2 | 169 | |||
AK297007.1 | - | 2 | 158 | |||
AK297760.1 | - | 2 | 257 | |||
AK314633.1 | - | 2 | 138 | |||
AK315948.1 | - | 2 | 118 | |||
AL049929.1 | - | 1 | 27 | |||
AY038990.1 | - | 2 | 190 | |||
AY429341.1 | - | 1 | 50 | |||
BC084541.1 | - | 2 | 167 | 6575732 |