Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282472.1 | NP_001269401.1 | 7 | 553 | ||
XM_006720651.3 | XP_006720714.1 | 7 | 573 | |||
XM_011521948.2 | XP_011520250.1 | 7 | 881 | |||
XM_011521950.2 | XP_011520252.1 | 7 | 879 | |||
GenBank mRNA | DB054802.1 | - | 7 | 442 |
Gene Symbol: GOLGA8H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282490.1 | NP_001269419.1 | 7 | 551 |
Gene Symbol: GOLGA8IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024074.1 | 6 | 754 | |||
GenBank mRNA | AK093104.1 | - | 6 | 754 |
Gene Symbol: GOLGA8K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282493.1 | NP_001269422.1 | 7 | 553 |
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 7 | 553 | ||
XM_005268284.4 | XP_005268341.1 | 7 | 1352 | |||
XM_005268285.4 | XP_005268342.1 | 7 | 1352 | |||
XM_006720654.3 | XP_006720717.1 | 7 | 496 | |||
XM_011521951.2 | XP_011520253.1 | 8 | 921 | |||
XM_011521952.2 | XP_011520254.1 | 7 | 1352 | |||
XM_011521953.2 | XP_011520255.1 | 7 | 1352 | |||
XM_011521954.2 | XP_011520256.1 | 7 | 486 |
Gene Symbol: GOLGA8T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033933.1 | 7 | 573 | |||
GenBank mRNA | AK310526.1 | - | 7 | 573 |