Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022782.3 | NP_073619.3 | 21 | 3397 | ||
NR_103517.1 | 21 | 3136 | ||||
XM_011537740.2 | XP_011536042.1 | 21 | 3301 | |||
XM_011537741.2 | XP_011536043.1 | 21 | 3301 | |||
XM_017018673.1 | XP_016874162.1 | 21 | 3395 | |||
XM_017018674.1 | XP_016874163.1 | 20 | 3199 | |||
XM_017018675.1 | XP_016874164.1 | 20 | 3203 | |||
XM_017018676.1 | XP_016874165.1 | 20 | 3145 | |||
XM_017018678.1 | XP_016874167.1 | 20 | 3121 | |||
XM_017018679.1 | XP_016874168.1 | 22 | 3334 | |||
XM_017018680.1 | XP_016874169.1 | 20 | 3243 | |||
XM_017018681.1 | XP_016874170.1 | 20 | 3156 | |||
GenBank mRNA | AK023016.1 | - | 16 | 2120 | ||
AK091181.1 | - | 17 | 2726 | |||
AK096541.1 | - | 21 | 3009 | |||
BC130440.1 | - | 17 | 2679 | 40146471 | ||
BC144017.1 | - | 16 | 2331 | |||
X98258.1 | - | 3 | 179 |