Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001560.2 | NP_001001560.1 | 3, 4 | 595 | ||
NM_001172687.1 | NP_001166158.1 | 3, 4 | 595 | |||
NM_001172688.1 | NP_001166159.1 | 4, 5 | 641 | |||
NM_013365.4 | NP_037497.1 | 3, 4 | 595 | |||
XM_005261518.1 | XP_005261575.1 | 3, 4 | 280 | |||
XM_005261520.1 | XP_005261577.1 | 3, 4 | 486 | |||
XM_006724229.1 | XP_006724292.1 | 4, 5 | 528 | |||
XM_017028760.1 | XP_016884249.1 | 4, 5 | 769 | |||
XM_017028762.1 | XP_016884251.1 | 4, 5 | 769 | |||
XM_017028763.1 | XP_016884252.1 | 3, 4 | 235 | |||
XM_017028764.1 | XP_016884253.1 | 3, 4 | 486 | |||
GenBank mRNA | AF190862.1 | - | 3, 4 | 223 | ||
AF218584.1 | - | 3, 4 | 212 | |||
AF233521.1 | - | 3, 4 | 230 | |||
AK075256.1 | - | 3, 4 | 573 | |||
AK122898.1 | - | 4, 5 | 641 | |||
AK225157.1 | - | 3, 4 | 229 | |||
AK290548.1 | - | 3, 4 | 234 | |||
AL110219.1 | - | 2, 3 | 119 | |||
BC044629.1 | - | 3, 4 | 224 | 5796877 |