Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018855.2 | NP_061343.2 | 10 | 1370 | ||
GenBank mRNA | AK140135.1 | - | 10 | 1348 | ||
AY692443.1 | - | 10 | 1369 | |||
BC003779.1 | - | 10 | 1285 | 3592695 | ||
U19859.1 | - | 10 | 1391 |