Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015889.2 | NP_001015889.1 | 3 | 735 | ||
NM_027139.5 | NP_081415.1 | 3 | 727 | |||
GenBank mRNA | AF305839.1 | - | 2 | 625 | ||
AK009322.1 | - | 1 | 475 | |||
BC042723.1 | - | 3 | 702 | 5006430 | ||
BC043028.1 | - | 2 | 559 | 5692221 | ||
BC096683.1 | - | 2 | 1475 | |||
BC108348.1 | - | 3 | 657 | 3994683 | ||
CN837441.1 | - | 1 | 376 |