Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054074.1 | NP_473415.1 | 2 | 208 | ||
GenBank mRNA | AB063109.1 | - | 2 | 208 | ||
BC119422.1 | - | 2 | 208 | 40038954 | ||
BC119423.2 | - | 2 | 208 | 40038955 | ||
BC141871.1 | - | 2 | 208 | 40038967 | ||
BC141872.1 | - | 2 | 208 | 40038968 |