Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010167.4 | NP_034297.2 | 11 | 1142 | ||
GenBank mRNA | AK041625.1 | - | 11 | 1143 | ||
BC120898.1 | - | 11 | 1126 | 40061597 | ||
BC120899.1 | - | 11 | 1126 | 40061599 | ||
Y17115.1 | - | 12 | 1253 |