Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030599.4 | NP_085102.4 | 3 | 276 | ||
GenBank mRNA | AF324826.1 | - | 3 | 252 | ||
AF338321.1 | - | 3 | 207 | |||
AF338322.1 | - | 3 | 207 | |||
AF342896.1 | - | 3 | 255 | |||
AF354259.1 | - | 213 | ||||
AF354260.1 | - | 213 | ||||
AK030702.1 | - | 3 | 225 | |||
AK041228.1 | - | 3 | 277 | |||
AK138766.1 | - | 3 | 275 | |||
AK169820.1 | - | 3 | 293 | |||
BC100580.1 | - | 3 | 263 | 30074482 | ||
BC132126.1 | - | 3 | 251 | 40130403 | ||
BC132128.1 | - | 3 | 251 | 40130405 | ||
BC145389.1 | - | 3 | 251 | 9053947 | ||
DQ237928.1 | - | 3 | 209 | |||
HQ713433.1 | - | 207 | ||||
M77677.1 | - | 3 | 252 | |||
X64719.1 | - | 180 |