Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008889.2 | NP_032915.2 | 4 | 879 | ||
GenBank mRNA | AK165792.1 | - | 4 | 493 | ||
BC082545.1 | - | 4 | 879 | 6820968 | ||
CF739461.1 | - | 4 | 483 | |||
X97490.1 | - | 4 | 942 |