Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145999.2 | NP_666111.1 | 13, 14 | 1514 | ||
GenBank mRNA | AY311390.1 | - | 13, 14 | 1514 | ||
BC023163.1 | - | 3, 4 | 284 | |||
BC023927.1 | - | 1, 2 | 318 | |||
BC029777.1 | - | 13, 14 | 1147 | 4950061 | ||
BC034062.1 | - | 13, 14 | 1169 | 4207654 |