Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166482.1 | NP_001159954.1 | 8 | 900 | ||
NM_172761.3 | NP_766349.1 | 8 | 994 | |||
GenBank mRNA | AK083317.1 | - | 8 | 934 | ||
AK141278.1 | - | 8 | 829 | |||
BC023782.1 | - | 8 | 971 | |||
BC026652.1 | - | 7 | 870 | |||
BC052066.1 | - | 7 | 2277 | 5708660 |