Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166482.1 | NP_001159954.1 | 4 | 358 | ||
NM_172761.3 | NP_766349.1 | 4 | 452 | |||
GenBank mRNA | AK083317.1 | - | 4 | 392 | ||
AK141278.1 | - | 4 | 287 | |||
BC023782.1 | - | 4 | 429 | |||
BC026652.1 | - | 3 | 328 | |||
BC052066.1 | - | 4 | 358 | 5708660 | ||
BQ444001.1 | - | 4 | 358 |