Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177164.3 | NP_796138.2 | 2 | 1081 | ||
GenBank mRNA | AB374230.1 | - | 1 | 277 | ||
AK080585.1 | - | 4 | 530 | |||
AK083856.1 | - | 2 | 1046 | |||
BC064448.1 | - | 2 | 894 | |||
BC094541.1 | - | 2 | 1081 | 6410144 | ||
EF552208.1 | - | 1 | 345 | |||
EF552209.1 | - | 1 | 280 | |||
EF552210.1 | - | 2 | 139 |