Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164679.1 | NP_001158151.1 | 15 | 2648 | ||
GenBank mRNA | AK173222.1 | - | 13 | 3636 | ||
BC027735.1 | - | 15 | 2550 | |||
BC027790.1 | - | 8 | 2122 | |||
BC042391.1 | - | 8 | 2843 | |||
BC053460.1 | - | 3 | 687 | |||
BC059855.1 | - | 6 | 1181 |