Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207573.1 | NP_997456.1 | 1 | 586 | ||
GenBank mRNA | BC106971.1 | - | 1 | 586 | 40041445 | |
BC106972.1 | - | 1 | 586 | 40041448 |
Gene Symbol: Olfr1381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146469.2 | NP_666680.2 | 1 | 586 | ||
GenBank mRNA | BC106974.1 | - | 1 | 586 |
Gene Symbol: Olfr1382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011790.1 | NP_001011790.1 | 1 | 586 | ||
GenBank mRNA | BC119435.1 | - | 1 | 586 | 40041463 |