Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006353.2 | NP_006344.1 | 2 | 459 | ||
XM_017010171.1 | XP_016865660.1 | 2 | 518 | |||
XM_017010172.1 | XP_016865661.1 | 3 | 582 | |||
XM_017010173.1 | XP_016865662.1 | 4 | 620 | |||
GenBank mRNA | AK311841.1 | - | 2 | 407 | ||
AU143224.1 | - | 2 | 459 | |||
BC001282.1 | - | 2 | 403 | 3452950 | ||
BT009824.1 | - | 1 | 221 | |||
U90549.1 | - | 1 | 912 |