Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027643.1 | NP_081919.1 | 7 | 824 | ||
GenBank mRNA | AK016504.1 | - | 7 | 785 | ||
AK033510.1 | - | 7 | 646 | |||
AK033896.1 | - | 7 | 655 | |||
AK035418.1 | - | 6 | 1542 | |||
AK162625.1 | - | 7 | 658 | |||
AK170548.1 | - | 7 | 650 | |||
AK202080.1 | - | 2 | 105 | |||
AK205460.1 | - | 1 | 57 | |||
AK209446.1 | - | 2 | 104 | |||
BC020020.1 | - | 7 | 675 | 3156924 | ||
BC049808.1 | - | 1 | 62 | |||
BC058212.1 | - | 2 | 483 |