Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20768
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
8
968
NM_001034955.1
NP_001030127.1
8
968
NM_001034956.1
NP_001030128.1
8
761
NM_001034957.1
NP_001030129.1
5
476
NM_001290294.1
NP_001277223.1
10
1155
NM_001290295.1
NP_001277224.1
8
690
NM_001290296.1
NP_001277225.1
7
941
NM_001290297.1
NP_001277226.1
6
503
NM_001290298.1
NP_001277227.1
6
572
NM_006434.2
NP_006425.2
5
476
NM_015385.3
NP_056200.1
9
1059
NM_024991.2
NP_079267.1
9
786
XM_006717589.1
XP_006717652.1
9
1081
XM_006717593.1
XP_006717656.1
7
641
XM_011539140.1
XP_011537442.1
11
1825
XM_011539150.1
XP_011537452.1
11
1825
XM_011539155.1
XP_011537457.1
9
1727
XM_011539167.1
XP_011537469.1
8
1010
XM_017015500.1
XP_016870989.1
11
1825
XM_017015501.1
XP_016870990.1
11
1825
XM_017015502.1
XP_016870991.1
11
1825
XM_017015503.1
XP_016870992.1
11
1825
XM_017015504.1
XP_016870993.1
9
739
XM_017015505.1
XP_016870994.1
11
1825
XM_017015506.1
XP_016870995.1
9
739
XM_017015507.1
XP_016870996.1
11
1825
XM_017015508.1
XP_016870997.1
9
739
XM_017015509.1
XP_016870998.1
8
668
XM_017015510.1
XP_016870999.1
11
1825
XM_017015511.1
XP_016871000.1
9
739
XM_017015512.1
XP_016871001.1
7
641
XM_017015513.1
XP_016871002.1
7
641
XM_017015514.1
XP_016871003.1
7
641
XM_017015515.1
XP_016871004.1
10
1108
XM_017015516.1
XP_016871005.1
9
1081
XM_017015517.1
XP_016871006.1
10
1108
XM_017015518.1
XP_016871007.1
9
1037
XM_017015519.1
XP_016871008.1
8
1010
XM_017015520.1
XP_016871009.1
10
1108
XM_017015521.1
XP_016871010.1
9
1037
XM_017015522.1
XP_016871011.1
9
1081
XM_017015523.1
XP_016871012.1
9
1037
XM_017015524.1
XP_016871013.1
8
1010
XM_017015525.1
XP_016871014.1
8
668
XM_017015526.1
XP_016871015.1
9
739
XM_017015527.1
XP_016871016.1
8
668
XM_017015528.1
XP_016871017.1
8
712
XM_017015529.1
XP_016871018.1
9
739
XM_017015530.1
XP_016871019.1
9
739
XM_017015531.1
XP_016871020.1
8
668
XM_017015532.1
XP_016871021.1
8
712
XM_017015533.1
XP_016871022.1
7
641
XM_017015534.1
XP_016871023.1
9
739
XM_017015535.1
XP_016871024.1
8
668
XM_017015536.1
XP_016871025.1
8
668
XM_017015537.1
XP_016871026.1
8
712
XM_017015538.1
XP_016871027.1
8
712
XM_017015539.1
XP_016871028.1
7
641
XM_017015540.1
XP_016871029.1
7
641
GenBank mRNA
AB037717.1
-
9
718
AF330623.1
-
5
458
AF356525.1
-
8
979
AF356526.1
-
8
979
AF356527.1
-
8
772
AJ489942.1
-
5
527
AK300407.1
-
8
643
AK304753.1
-
9
1702
AL117472.1
-
9
1017
AM260536.1
-
10
1113
BC042612.1
-
7
1083
BC152463.1
-
9
718
8862599
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0