Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286474.1 | NP_001273403.1 | 2 | 274 | ||
NM_001286475.1 | NP_001273404.1 | 2 | 274 | |||
NM_006781.4 | NP_006772.3 | 2 | 274 | |||
XM_017010185.1 | XP_016865674.1 | 2 | 266 | |||
GenBank mRNA | AK225742.1 | - | 2 | 265 | ||
AK302176.1 | - | 2 | 234 | |||
AK310230.1 | - | 2 | 263 | |||
BC034774.1 | - | 2 | 241 | 5169157 | ||
DC399589.1 | - | 2 | 275 | |||
U60665.1 | - | 2 | 304 |