siRNA Details
Gene Symbol: Hoxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010452.3 | NP_034582.1 | 4 | 794 | ||
GenBank mRNA | BC096612.1 | - | 3 | 770 | 6476230 | |
U56399.1 | - | 1 | 303 | |||
Y11717.1 | - | 4 | 844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
