Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007018.4 | NP_008949.4 | 3 | 474 | ||
XM_005251679.3 | XP_005251736.1 | 5 | 746 | |||
XM_006716940.2 | XP_006717003.1 | 5 | 746 | |||
XM_006716944.3 | XP_006717007.1 | 5 | 746 | |||
XM_011518166.1 | XP_011516468.1 | 5 | 746 | |||
XM_011518167.1 | XP_011516469.1 | 4 | 587 | |||
XM_011518168.1 | XP_011516470.1 | 5 | 746 | |||
XM_011518169.2 | XP_011516471.1 | 5 | 741 | |||
XM_011518170.1 | XP_011516472.1 | 5 | 746 | |||
XM_011518172.2 | XP_011516474.1 | 5 | 746 | |||
XM_017014223.1 | XP_016869712.1 | 5 | 746 | |||
XM_017014224.1 | XP_016869713.1 | 5 | 746 | |||
XM_017014227.1 | XP_016869716.1 | 4 | 764 | |||
XM_017014229.1 | XP_016869718.1 | 5 | 746 | |||
XM_017014230.1 | XP_016869719.1 | 5 | 746 | |||
GenBank mRNA | AF513978.1 | - | 3 | 443 | ||
AK307387.1 | - | 5 | 751 | |||
BC089415.1 | - | 5 | 703 | |||
BC137284.1 | - | 3 | 467 | 9021281 | ||
BC137286.1 | - | 3 | 466 | 9021283 |