Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136162.1 | NP_001129634.1 | 17 | 1976 | ||
XM_006255802.2 | XP_006255864.1 | 19 | 2144 | |||
XM_006255803.2 | XP_006255865.1 | 19 | 2141 | |||
XM_006255805.2 | XP_006255867.1 | 17 | 1982 | |||
GenBank mRNA | AY724473.1 | - | 7 | 613 | ||
BC098840.1 | - | 3 | 260 | |||
BC105871.1 | - | 9 | 961 |