Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134833.2 | NP_001128305.1 | 1 | 369 | ||
GenBank mRNA | BC167063.1 | - | 1 | 369 | 7453194 |
Gene Symbol: LOC100911776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748922.3 | XP_003748970.1 | 1 | 1909 |