Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301065.1 | NP_001287994.1 | 5 | 556 | ||
NM_007180.2 | NP_009111.2 | 5 | 556 | |||
GenBank mRNA | AB000824.1 | - | 5 | 566 | ||
AK223140.1 | - | 5 | 530 | |||
AK223143.1 | - | 5 | 534 | |||
AK309565.1 | - | 5 | 530 | |||
BC109206.2 | - | 5 | 550 | 40006501 |