Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005871.1 | NP_001005871.1 | 11 | 2082 | ||
XM_008769446.1 | XP_008767668.1 | 12 | 2465 | |||
XM_008769447.1 | XP_008767669.1 | 12 | 3102 | |||
XM_008769448.1 | XP_008767670.1 | 11 | 2429 | |||
XM_008769449.1 | XP_008767671.1 | 12 | 2466 | |||
XM_008769450.1 | XP_008767672.1 | 11 | 2430 | |||
GenBank mRNA | U15408.1 | - | 11 | 2082 |