Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247063.2 | XP_006247125.1 | 9 | 2443 | ||
XM_008768157.1 | XP_008766379.1 | 9 | 1460 | |||
XM_008768158.1 | XP_008766380.1 | 9 | 2443 | |||
XM_008768159.1 | XP_008766381.1 | 9 | 2566 | |||
XM_340869.5 | XP_340870.3 | 9 | 1343 |