Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017414.3 | NP_059110.2 | 5 | 779 | ||
XM_006724074.2 | XP_006724137.1 | 5 | 850 | |||
GenBank mRNA | AF176642.2 | - | 4 | 461 | ||
AJ243526.1 | - | 5 | 644 | |||
AK313385.1 | - | 5 | 600 | |||
AL136690.1 | - | 5 | 600 | |||
BC014896.1 | - | 5 | 585 | 3910720 | ||
BI752849.1 | - | 5 | 673 | |||
BT006835.1 | - | 4 | 441 | |||
CR457216.1 | - | 4 | 441 | |||
CT841507.1 | - | 4 | 507 |
Gene Symbol: USP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029164.1 | XP_016884653.1 | 5 | 375 | ||
GenBank mRNA | AJ583822.1 | - | 4 | 348 | ||
AJ586979.1 | - | 3 | 285 | |||
AK303463.1 | - | 2 | 194 |