Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNA SIRNA Assays ›
Gene Symbol:
DTNA
Gene Name:
dystrobrevin alpha
Gene Aliases:
D18S892E, DRP3, DTN, DTN-A, LVNC1
Chromosome Location:
Chr.18: 34493290 - 34891844 on Build GRCh38
UniGene ID:
Hs.643454
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223448
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128175.1
NP_001121647.1
10
1334
NM_001198938.1
NP_001185867.1
10
1334
NM_001198939.1
NP_001185868.1
10
1334
NM_001198940.1
NP_001185869.1
10
1334
NM_001198941.1
NP_001185870.1
9
1202
NM_001198942.1
NP_001185871.1
2
476
NM_001198943.1
NP_001185872.1
2
480
NM_001198944.1
NP_001185873.1
2
476
NM_001390.4
NP_001381.2
8
988
NM_001391.5
NP_001382.2
8
988
NM_001392.4
NP_001383.2
10
1338
NM_032975.3
NP_116757.2
10
1338
NM_032978.6
NP_116760.2
9
1202
NM_032979.4
NP_116761.2
10
1338
NM_032980.3
NP_116762.2
2
421
NM_032981.4
NP_116763.1
2
421
XM_005258221.4
XP_005258278.1
10
1333
XM_011525853.2
XP_011524155.1
10
1333
XM_017025575.1
XP_016881064.1
9
1202
XM_017025576.1
XP_016881065.1
10
1327
XM_017025577.1
XP_016881066.1
9
1202
XM_017025578.1
XP_016881067.1
10
1327
XM_017025579.1
XP_016881068.1
9
1202
XM_017025580.1
XP_016881069.1
10
1333
XM_017025581.1
XP_016881070.1
10
1327
XM_017025582.1
XP_016881071.1
9
1202
XM_017025583.1
XP_016881072.1
10
1333
XM_017025584.1
XP_016881073.1
10
1327
XM_017025585.1
XP_016881074.1
10
1327
XM_017025586.1
XP_016881075.1
10
1327
XM_017025587.1
XP_016881076.1
10
1327
XM_017025588.1
XP_016881077.1
9
1202
XM_017025589.1
XP_016881078.1
10
1333
XM_017025590.1
XP_016881079.1
10
1327
XM_017025591.1
XP_016881080.1
9
1202
XM_017025592.1
XP_016881081.1
10
1333
XM_017025593.1
XP_016881082.1
10
1327
XM_017025594.1
XP_016881083.1
9
1202
XM_017025595.1
XP_016881084.1
10
1333
XM_017025596.1
XP_016881085.1
10
1333
XM_017025597.1
XP_016881086.1
10
1327
XM_017025598.1
XP_016881087.1
9
1202
XM_017025599.1
XP_016881088.1
2
395
XM_017025600.1
XP_016881089.1
9
1202
XM_017025601.1
XP_016881090.1
10
1327
XM_017025602.1
XP_016881091.1
9
1202
GenBank mRNA
AB209102.1
-
10
1287
AJ009668.1
-
10
1246
AK291156.1
-
9
1137
AK295732.1
-
2
379
AK295789.1
-
2
367
AK296450.1
-
2
481
AK308410.1
-
10
1314
AK315215.1
-
2
312
AL833285.1
-
10
1294
BC005300.1
-
10
1294
3933795
BG705393.1
-
2
404
BI668892.1
-
2
398
BT006937.1
-
8
987
DC327958.1
-
2
421
DN991561.1
-
2
483
FJ535564.1
-
2
180
FJ535565.1
-
2
180
U26742.1
-
10
1311
U26743.1
-
2
368
U26744.1
-
9
1045
U46744.1
-
9
1051
U46745.1
-
9
1258
U46746.1
-
2
371
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001285813, NM_001285817, NM_001285808, NM_001285807, NM_001285811, and others.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
striated muscle contraction
signal transduction
chemical synaptic transmission
neuromuscular synaptic transmission
protein binding
zinc ion binding
PDZ domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0