Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015629.3 | NP_056444.3 | 7 | 1050 | ||
XM_006723137.3 | XP_006723200.1 | 7 | 960 | |||
GenBank mRNA | AB593024.1 | - | 8 | 1969 | ||
AB593025.1 | - | 7 | 706 | |||
AF308303.1 | - | 6 | 525 | |||
AK098547.1 | - | 7 | 689 | |||
AK301709.1 | - | 7 | 715 | |||
AL050369.1 | - | 7 | 689 | |||
AY040822.1 | - | 7 | 1011 | |||
BC117389.1 | - | 7 | 974 | 40125940 |