Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000170.2 | NP_000161.2 | 16 | 2089 | ||
GenBank mRNA | AK308470.1 | - | 2 | 331 | ||
AK310534.1 | - | 12 | 1464 | |||
AK314156.1 | - | 16 | 2091 | |||
BC111993.1 | - | 16 | 1962 | 8327461 | ||
BC111995.1 | - | 16 | 1962 | 8327463 | ||
D90239.1 | - | 16 | 2046 | |||
M63635.1 | - | 16 | 1977 | |||
M64590.1 | - | 16 | 2046 |