Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002254.6 | NP_002245.4 | 7 | 2923 | ||
XM_005264299.2 | XP_005264356.2 | 7 | 2920 | |||
GenBank mRNA | AF018164.1 | - | 7 | 2417 | ||
AF035621.1 | - | 7 | 2419 | |||
AK293738.1 | - | 8 | 2187 | |||
AK315324.1 | - | 8 | 2947 | |||
BC018932.2 | - | 8 | 899 | |||
BC042486.1 | - | 8 | 2830 | 3866587 | ||
BC092406.1 | - | 7 | 2808 | 6504352 | ||
BC130423.1 | - | 7 | 2494 | 40146454 | ||
BC132783.1 | - | 7 | 2494 | 40146805 | ||
BC144223.1 | - | 7 | 2488 | 9052743 | ||
BX571741.1 | - | 7 | 2846 |