Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224348
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
8
786
NM_001077352.1
NP_001070820.1
6
684
NM_001308044.1
NP_001294973.1
6
625
NM_018107.4
NP_060577.3
7
738
XM_011536890.2
XP_011535192.1
9
869
XM_011536892.2
XP_011535194.1
8
820
XM_011536893.2
XP_011535195.1
8
821
XM_011536894.2
XP_011535196.1
8
815
XM_011536895.2
XP_011535197.1
7
791
XM_011536896.2
XP_011535198.1
7
767
XM_011536897.2
XP_011535199.1
7
1153
XM_011536900.1
XP_011535202.1
7
729
XM_011536902.1
XP_011535204.1
7
635
XM_011536903.1
XP_011535205.1
5
511
XM_011536904.1
XP_011535206.1
6
568
XM_011536905.1
XP_011535207.1
5
566
XM_011536906.1
XP_011535208.1
4
457
XM_017021398.1
XP_016876887.1
8
823
XM_017021399.1
XP_016876888.1
7
770
XM_017021400.1
XP_016876889.1
7
688
XM_017021401.1
XP_016876890.1
8
783
XM_017021402.1
XP_016876891.1
8
694
XM_017021403.1
XP_016876892.1
7
712
XM_017021404.1
XP_016876893.1
7
662
XM_017021405.1
XP_016876894.1
7
729
XM_017021406.1
XP_016876895.1
8
1201
XM_017021407.1
XP_016876896.1
8
710
XM_017021408.1
XP_016876897.1
6
646
XM_017021409.1
XP_016876898.1
7
1147
XM_017021410.1
XP_016876899.1
6
668
XM_017021411.1
XP_016876900.1
6
1099
XM_017021412.1
XP_016876901.1
8
821
XM_017021413.1
XP_016876902.1
7
597
XM_017021414.1
XP_016876903.1
7
635
XM_017021415.1
XP_016876904.1
6
616
XM_017021416.1
XP_016876905.1
5
511
XM_017021417.1
XP_016876906.1
6
568
XM_017021418.1
XP_016876907.1
5
566
XM_017021419.1
XP_016876908.1
4
457
XM_017021420.1
XP_016876909.1
5
986
GenBank mRNA
AF087905.1
-
7
689
AK308055.1
-
7
701
BC024208.2
-
7
675
2960467
BX161440.1
-
8
719
CK000110.1
-
7
711
CK003276.1
-
6
616
CR457257.1
-
6
539
KU178687.1
-
6
539
KU178688.1
-
7
587
KU178689.1
-
5
485
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0