Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUF60 SIRNA Assays ›
Gene Symbol:
PUF60
Gene Name:
poly(U) binding splicing factor 60
Gene Aliases:
FIR, RoBPI, SIAHBP1, VRJS
Chromosome Location:
Chr.8: 143816344 - 143829386 on Build GRCh38
UniGene ID:
Hs.521924
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22447
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUF60
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136033.2
NP_001129505.1
6
499
NM_001271096.1
NP_001258025.1
5
426
NM_001271097.1
NP_001258026.1
4
342
NM_001271098.1
NP_001258027.1
6
477
NM_001271099.1
NP_001258028.1
5
393
NM_001271100.1
NP_001258029.1
5
448
NM_014281.4
NP_055096.2
5
429
NM_078480.2
NP_510965.1
6
480
XM_011516929.2
XP_011515231.1
7
679
XM_011516930.2
XP_011515232.1
6
530
XM_017013234.1
XP_016868723.1
6
2012
XM_017013235.1
XP_016868724.1
6
582
XM_017013236.1
XP_016868725.1
5
1961
XM_017013237.1
XP_016868726.1
7
741
XM_017013238.1
XP_016868727.1
7
541
XM_017013239.1
XP_016868728.1
6
507
XM_017013240.1
XP_016868729.1
5
456
GenBank mRNA
AF114818.1
-
6
461
AF190744.1
-
6
389
AF217197.2
-
5
356
AK055941.1
-
5
1083
AL522753.3
-
6
427
BC008875.2
-
5
395
4138778
BC009734.1
-
5
378
3877563
BC011265.1
-
6
416
4155549
BC011979.1
-
4
295
3863660
BC035435.2
-
5
380
BG771821.1
-
5
447
BX418925.2
-
5
378
CN371576.1
-
5
396
CR457143.1
-
6
397
CT000277.1
-
5
361
CX163878.1
-
4
300
U51586.1
-
4
343
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
apoptotic process
RNA splicing
cell-cell adhesion
nucleotide binding
DNA binding
protein binding
identical protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kt7sl:80/10.239.164.98:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0