Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172732.1 | NP_001166203.1 | 2 | 333 | ||
NM_001172739.1 | NP_001166210.1 | 3 | 527 | |||
NM_001172743.1 | NP_001166214.1 | 3 | 530 | |||
NM_021785.4 | NP_068557.3 | 2 | 333 | |||
NR_033348.1 | 3 | 606 | ||||
NR_033349.1 | 2 | 412 | ||||
XM_006724459.2 | XP_006724522.1 | 4 | 682 | |||
XM_006724460.1 | XP_006724523.1 | 3 | 430 | |||
XM_011545439.2 | XP_011543741.1 | 4 | 711 | |||
XM_011545440.2 | XP_011543742.1 | 4 | 714 | |||
XM_011545441.2 | XP_011543743.1 | 3 | 517 | |||
GenBank mRNA | AK056214.1 | - | 3 | 426 | ||
AK298873.1 | - | 2 | 294 | |||
AK314631.1 | - | 2 | 296 | |||
BC027937.1 | - | 3 | 486 | 5201821 | ||
DA377448.1 | - | 2 | 357 | |||
DA648693.1 | - | 2 | 334 | |||
DC420191.1 | - | 3 | 491 | |||
DR005700.1 | - | 2 | 418 |