Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080868.2 | NP_543144.1 | 1 | 409 | ||
NR_026546.1 | 1 | 409 | ||||
GenBank mRNA | AF403035.1 | - | 1 | 269 | ||
AF429968.1 | - | 1 | 386 | |||
AK098606.1 | - | 1 | 409 | |||
BC022517.1 | - | 1 | 342 | |||
BC036573.2 | - | 1 | 387 | 5297837 |