Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225770
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
18
3380
NM_152586.3
NP_689799.3
18
3478
NR_135249.1
19
3865
NR_135250.1
14
2860
XM_005269582.2
XP_005269639.1
17
6556
XM_011539368.2
XP_011537670.1
18
6622
XM_017015759.1
XP_016871248.1
20
6051
XM_017015760.1
XP_016871249.1
20
4953
XM_017015761.1
XP_016871250.1
20
5791
XM_017015762.1
XP_016871251.1
20
4961
XM_017015763.1
XP_016871252.1
20
4255
XM_017015764.1
XP_016871253.1
20
4318
XM_017015765.1
XP_016871254.1
21
4571
XM_017015766.1
XP_016871255.1
20
6177
XM_017015767.1
XP_016871256.1
20
5836
XM_017015768.1
XP_016871257.1
20
4007
XM_017015769.1
XP_016871258.1
20
3615
XM_017015770.1
XP_016871259.1
20
4276
XM_017015771.1
XP_016871260.1
19
6793
XM_017015772.1
XP_016871261.1
19
6799
XM_017015773.1
XP_016871262.1
18
6733
XM_017015774.1
XP_016871263.1
19
5719
XM_017015775.1
XP_016871264.1
20
5597
XM_017015776.1
XP_016871265.1
19
6102
XM_017015777.1
XP_016871266.1
19
5761
XM_017015778.1
XP_016871267.1
18
6679
XM_017015779.1
XP_016871268.1
19
6799
XM_017015780.1
XP_016871269.1
19
6799
XM_017015781.1
XP_016871270.1
18
6628
XM_017015782.1
XP_016871271.1
19
4379
XM_017015783.1
XP_016871272.1
19
5426
XM_017015784.1
XP_016871273.1
18
3372
XM_017015785.1
XP_016871274.1
18
6679
XM_017015786.1
XP_016871275.1
18
3869
GenBank mRNA
AK094637.1
-
1
572
AK126497.1
-
6
1833
AK127213.1
-
5
1413
AK127267.1
-
14
2860
AK303633.1
-
3
1071
AL832102.1
-
1
518
BC026037.1
-
1
326
BC064967.1
-
1
514
BC130633.1
-
3
1062
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0