Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153368.2 | NP_699199.2 | 2 | 348 | ||
GenBank mRNA | AJ414564.1 | - | 2 | 190 | ||
AK074504.1 | - | 2 | 348 |