Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012478.3 | NP_036610.2 | 1 | 165 | ||
XM_011524580.2 | XP_011522882.1 | 1 | 86 | |||
GenBank mRNA | AF087866.1 | - | 1 | 69 | ||
AK057881.1 | - | 1 | 70 | |||
AK293214.1 | - | 1 | 54 | |||
AK294082.1 | - | 1 | 80 | |||
AK294155.1 | - | 1 | 80 | |||
AK300875.1 | - | 1 | 80 | |||
BC007452.1 | - | 1 | 68 | 3140484 | ||
BC010616.1 | - | 1 | 78 | 4177319 | ||
DA278794.1 | - | 1 | 165 |