Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278295.1 | NP_001265224.1 | 5 | 376 | ||
NM_001278296.1 | NP_001265225.1 | 4 | 327 | |||
NM_152389.3 | NP_689602.2 | 4 | 327 | |||
NM_194302.3 | NP_919278.2 | 5 | 482 | |||
XM_011510903.1 | XP_011509205.1 | 5 | 543 | |||
XM_011510904.2 | XP_011509206.1 | 5 | 2127 | |||
XM_011510908.1 | XP_011509210.1 | 5 | 449 | |||
XM_011510909.1 | XP_011509211.1 | 5 | 489 | |||
XM_011510910.1 | XP_011509212.1 | 5 | 543 | |||
XM_011510911.1 | XP_011509213.1 | 5 | 394 | |||
XM_011510912.1 | XP_011509214.1 | 5 | 437 | |||
XM_011510914.1 | XP_011509216.1 | 4 | 334 | |||
XM_011510915.1 | XP_011509217.1 | 4 | 296 | |||
XM_011510916.1 | XP_011509218.1 | 5 | 543 | |||
XM_011510919.1 | XP_011509221.1 | 5 | 543 | |||
XM_017003752.1 | XP_016859241.1 | 6 | 610 | |||
XM_017003753.1 | XP_016859242.1 | 5 | 1967 | |||
XM_017003754.1 | XP_016859243.1 | 4 | 388 | |||
XM_017003755.1 | XP_016859244.1 | 4 | 1972 | |||
GenBank mRNA | AK127189.1 | - | 5 | 447 | ||
AK302678.1 | - | 4 | 292 | |||
AK302688.1 | - | 5 | 341 | |||
BC031585.1 | - | 4 | 299 | |||
BC047637.1 | - | 5 | 352 | |||
BG723301.1 | - | 5 | 357 | |||
BX649039.1 | - | 2 | 854 | |||
DB070903.1 | - | 5 | 449 |