Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002370.3 | NP_002361.1 | 5 | 522 | ||
GenBank mRNA | AF035940.1 | - | 5 | 425 | ||
AF067173.1 | - | 5 | 448 | |||
AF086246.1 | - | 3 | 161 | |||
AK297895.1 | - | 4 | 328 | |||
AK312113.1 | - | 5 | 458 | |||
BC018211.1 | - | 5 | 434 | 3861094 |