Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317770.1 | NP_001304699.1 | 3 | 447 | ||
NM_017592.2 | NP_060062.1 | 3 | 447 | |||
NR_133915.1 | 2 | 388 | ||||
GenBank mRNA | AK000411.1 | - | 3 | 357 | ||
AK090877.1 | - | 2 | 279 | |||
AK124629.1 | - | 4 | 310 | |||
AK294667.1 | - | 4 | 269 | |||
AK297772.1 | - | 3 | 338 | |||
AK298014.1 | - | 3 | 357 | |||
AK300563.1 | - | 3 | 439 | |||
AK316010.1 | - | 4 | 278 | |||
AL137304.1 | - | 3 | 362 | |||
AY729650.1 | - | 3 | 427 | |||
BC019015.2 | - | 3 | 354 | 4129619 | ||
DA224505.1 | - | 2 | 389 |